CAS RN 67515-74-6
IUPAC Name:
[4-fluoro-3-(trifluoromethyl)phenyl]methanaminium
Synonyms:
[4-fluoro-3-(trifluoromethyl)phenyl]methanaminium
Molecular Weight (g/mol):
194.15
Molecular Formula:
C8H8F4N
InChi Key:
HZDVQEUISWBXPV-UHFFFAOYSA-O
SMILES:
[NH3+]CC1=CC=C(F)C(=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Fluoro-3-(trifluoromethyl)benzylamine, 94%, Thermo Scientific™
CAS: 67515-74-6 Molecular Formula: C8H8F4N Molecular Weight (g/mol): 194.15 MDL Number: MFCD00070814 InChI Key: HZDVQEUISWBXPV-UHFFFAOYSA-O Synonym: 4-fluoro-3-trifluoromethyl benzylamine,4-fluoro-3-trifluoromethyl phenyl methanamine,4-fluoro-3-trifluoromethyl benzyl amine,4-fluoro-3-trifluoromethylbenzylamine,4-fluoro-3-trifluoromethyl-benzylamine,benzenemethanamine, 4-fluoro-3-trifluoromethyl,4-fluoro-3-trifluoromethyl phenyl methylamine,1-4-fluoro-3-trifluoromethyl phenyl methanamine,rarechem al bw 0670 PubChem CID: 522270 IUPAC Name: [4-fluoro-3-(trifluoromethyl)phenyl]methanamine SMILES: [NH3+]CC1=CC=C(F)C(=C1)C(F)(F)F