accessibility menu, dialog, popup

UserName

CAS RN 6639-82-3

CH3ONN

CAS RN 6639-82-3

IUPAC Name: 6-methoxyquinoxaline
Synonyms: 6-methoxyquinoxaline
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: BURIFIXTNVTJJN-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=CC=NC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results