CAS RN 66376-36-1
IUPAC Name:
(4-amino-1-hydroxy-1-phosphonobutyl)phosphonic acid
Synonyms:
alendronate alendronic acid monosodium salt trihydrate alendronate sodium aminohydroxybutane bisphosphonate acide alendronique alendronate sodium hydrate sodium, alendronate alendronate sodium - chemical alendronate monosodium salt, trihydrate aminohydroxybutylidene biphosphonate monosodium salt trihydrate
Molecular Weight (g/mol):
249.1
Molecular Formula:
C4H13NO7P2
InChi Key:
OGSPWJRAVKPPFI-UHFFFAOYSA-N
SMILES:
NCCCC(O)(P(O)(O)=O)P(O)(O)=O
1
–
1
of
1
results
1
Alendronate Acid, TRC
CAS: 66376-36-1 Molecular Formula: C4H13NO7P2 Molecular Weight (g/mol): 249.1 Synonym: P,P'-(4-Amino-1-hydroxybutylidene)bis[phosphonic Acid],4-Amino-1-hydroxybutane-1,1-bisphosphonic Acid,4-Amino-1-hydroxybutane-1,1-diphosphonate,4-Amino-1-hydroxybutane-1,1-diphosphonic Acid,4-Amino-1-hydroxybutane-1,1-diyldiphosphonic Acid,4-Amino-1-hydroxybutylidene-1,1-bis(phosphonic Acid),ABDP,Alendronic Acid(4-amino-1-hydroxybutylidene)bis-,BPH 1,Marvil,(4-Amino-1-hydroxybutylidene)bis-phosphonic Acid IUPAC Name: (4-amino-1-hydroxy-1-phosphonobutyl)phosphonic acid SMILES: C(CC(O)(P(=O)(O)O)P(=O)(O)O)CN