accessibility menu, dialog, popup

UserName

CAS RN 66298-09-7

CH3HNSHNNH2CH3

CAS RN 66298-09-7

IUPAC Name: 3-amino-1-(2,4-dimethylphenyl)thiourea
Synonyms: 3-amino-1-(2,4-dimethylphenyl)thiourea
Molecular Weight (g/mol): 195.28
Molecular Formula: C9H13N3S
InChi Key: YTRJQDFRZLKDHZ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(NC(=S)NN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results