CAS RN 662163-81-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Importazole, MedChemExpress
MedChemExpress Importazole is a small molecule inhibitor of the nuclear transport receptor importin-β.