CAS RN 66121-69-5
CAS RN 66121-69-5
IUPAC Name:
1-methyl-1H-imidazole-4-carbonitrile
Synonyms:
1-methylimidazole-4-carbonitrile
Molecular Weight (g/mol):
107.12
Molecular Formula:
C5H5N3
InChi Key:
LAKVILSENFFPRR-UHFFFAOYSA-N
SMILES:
CN1C=NC(=C1)C#N
1
–
1
of
1
results
1
1-Methyl-1H-imidazole-4-carbonitrile, 97%, Thermo Scientific™
CAS: 66121-69-5 Molecular Formula: C5H5N3 Molecular Weight (g/mol): 107.116 MDL Number: MFCD06411537 InChI Key: LAKVILSENFFPRR-UHFFFAOYSA-N PubChem CID: 2795115 IUPAC Name: 1-methylimidazole-4-carbonitrile SMILES: CN1C=C(N=C1)C#N