accessibility menu, dialog, popup

UserName

CAS RN 6601-20-3

IUPAC Name: 1,3-bis(prop-2-en-1-yl)thiourea
Synonyms: 1,3-bis(prop-2-en-1-yl)thiourea
Molecular Weight (g/mol): 156.25
Molecular Formula: C7H12N2S
InChi Key: DZZWKUMHMSNBSG-UHFFFAOYSA-N
SMILES: C=CCNC(=S)NCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results