CAS RN 65032-27-1
CAS RN 65032-27-1
IUPAC Name:
magnesium(2+) acetylene chloride
Synonyms:
magnesium(2+) acetylene chloride
Molecular Weight (g/mol):
84.79
Molecular Formula:
C2HClMg
InChi Key:
GWGVDNZFTPIGDY-UHFFFAOYSA-M
SMILES:
[Mg++].[Cl-].C#[C-]
1
–
1
of
1
results
1
Ethynylmagnesium chloride, 0.5M solution in THF/Toluene, AcroSeal™
CAS: 65032-27-1 Molecular Formula: C2HClMg Molecular Weight (g/mol): 84.79 MDL Number: MFCD00075343 InChI Key: YEWNLGTVYUADOR-UHFFFAOYSA-M Synonym: ethynylmagnesium chloride,ethynylmagnesiumchloride,ethynylchloromagnesium,ethynyl magnesium chloride,ethynyl-magnesium chloride,chloro ethynyl magnesium,yewnlgtvyuador-uhfffaoysa-m,ethynylmagnesium chloride solution, 0.5 m in thf PubChem CID: 2734899 IUPAC Name: chloro(ethynyl)magnesium SMILES: Cl[Mg]C#C