CAS RN 6481-48-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methylhistamine Dihydrochloride, TRC
CAS: 6481-48-7 Molecular Formula: C6 H11 N3 . 2HCl Molecular Weight (g/mol): 198.09352 Synonym: 4-(2-Aminoethyl)-1-methyl-imidazole Dihydrochloride,1-Methyl-1H-imidazole-4-ethanamine Dihydrochloride; IUPAC Name: 2-(1-methylimidazol-4-yl)ethanamine;dihydrochloride SMILES: Cl.Cl.Cn1cnc(CCN)c1