accessibility menu, dialog, popup

UserName

CAS RN 647855-21-8

H3CONH2OFFF

CAS RN 647855-21-8

IUPAC Name: 4-methoxy-3-(trifluoromethoxy)aniline
Synonyms: 4-methoxy-3-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 207.15
Molecular Formula: C8H8F3NO2
InChi Key: SUHKHUKMTRYZOA-UHFFFAOYSA-N
SMILES: COC1=C(OC(F)(F)F)C=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results