accessibility menu, dialog, popup

UserName

CAS RN 64017-81-8

NH2OH2NHCl

CAS RN 64017-81-8

IUPAC Name: 3-aminopropanamide hydrochloride
Synonyms: 3-aminopropionamide hydrochloride
Molecular Weight (g/mol): 124.57
Molecular Formula: C3H9ClN2O
InChi Key: GAGJMOQGABUOBK-UHFFFAOYSA-N
SMILES: Cl.NCCC(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)