CAS RN 64-04-0
CAS RN 64-04-0
IUPAC Name:
2-phenylethan-1-amine
Synonyms:
2-phenylethylamine β-phenethylamine
Molecular Weight (g/mol):
121.18
Molecular Formula:
C8H11N
InChi Key:
BHHGXPLMPWCGHP-UHFFFAOYSA-N
SMILES:
NCCC1=CC=CC=C1
1
–
1
of
1
results
1
2-Phenylethylamine, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality