CAS RN 6388-47-2
CAS RN 6388-47-2
IUPAC Name:
2-amino-3-chlorobenzoic acid
Synonyms:
2-amino-3-chlorobenzoic acid
Molecular Weight (g/mol):
171.58
Molecular Formula:
C7H6ClNO2
InChi Key:
LWUAMROXVQLJKA-UHFFFAOYSA-N
SMILES:
NC1=C(Cl)C=CC=C1C(O)=O