accessibility menu, dialog, popup

UserName

CAS RN 63843-42-5

CH3OHN

CAS RN 63843-42-5

IUPAC Name: 4-(2-methylphenoxy)piperidine
Synonyms: 4-(2-methylphenoxy)piperidine
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: MLLZZLRYRWFFRQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OC1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results