CAS RN 638213-98-6
CAS RN 638213-98-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AGL-2263, MedChemExpress
MedChemExpress AGL-2263 is an insulin receptor and insulin-like growth factor (IGF) receptor inhibitor.