accessibility menu, dialog, popup

UserName

CAS RN 638156-11-3

ONNHOOCl

CAS RN 638156-11-3

IUPAC Name: 5-(3-chlorophenyl)-N-{4-[(morpholin-4-yl)methyl]phenyl}furan-2-carboxamide
Synonyms: 5-(3-chlorophenyl)-N-[4-(morpholin-4-ylmethyl)phenyl]furan-2-carboxamide
Molecular Weight (g/mol): 396.87
Molecular Formula: C22H21ClN2O3
InChi Key: XQJWTJLJEYIUDZ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C1=CC=C(O1)C(=O)NC1=CC=C(CN2CCOCC2)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results