accessibility menu, dialog, popup

UserName

CAS RN 638-53-9

H3COHO

CAS RN 638-53-9

IUPAC Name: tridecanoic acid
Synonyms: N-tridecanoic acid
Molecular Weight (g/mol): 214.35
Molecular Formula: C13H26O2
InChi Key: SZHOJFHSIKHZHA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC(O)=O
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)