accessibility menu, dialog, popup

UserName

CAS RN 638-36-8

H3CCH3CH3CH3CH3CH3

CAS RN 638-36-8

IUPAC Name: 2,6,10,14-tetramethylhexadecane
Synonyms: phytane phytan
Molecular Weight (g/mol): 282.56
Molecular Formula: C20H42
InChi Key: GGYKPYDKXLHNTI-UHFFFAOYNA-N
SMILES: CCC(C)CCCC(C)CCCC(C)CCCC(C)C
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results