CAS RN 638-08-4
CAS RN 638-08-4
IUPAC Name:
octadecanoyl octadecanoate
Synonyms:
octadecanoic acid, anhydride
Molecular Weight (g/mol):
550.95
Molecular Formula:
C36H70O3
InChi Key:
WVJVHUWVQNLPCR-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC