accessibility menu, dialog, popup

UserName

CAS RN 63675-74-1

H3COSOCH3

CAS RN 63675-74-1

IUPAC Name: 6-methoxy-2-(4-methoxyphenyl)-1-benzothiophene
Synonyms: 6-methoxy-2-(4-methoxyphenyl)-1-benzothiophene
Molecular Weight (g/mol): 270.35
Molecular Formula: C16H14O2S
InChi Key: HRWAGCVMOGWQJF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC2=CC=C(OC)C=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results