accessibility menu, dialog, popup

UserName

CH3(R)(S)H3CNHNOClSNH2OO

CAS RN 636-54-4

IUPAC Name: 4-chloro-N-[(2R,6S)-2,6-dimethylpiperidin-1-yl]-3-sulfamoylbenzamide
Synonyms: clopamide
Molecular Weight (g/mol): 345.84
Molecular Formula: C14H20ClN3O3S
InChi Key: LBXHRAWDUMTPSE-AOOOYVTPSA-N
SMILES: C[C@H]1CCC[C@@H](C)N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results