CAS RN 63546-27-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
L-Alanine-3,3,3-d3, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
L-Alanine-d3, TRC
CAS: 63546-27-0 Molecular Formula: C3 D3 H4 N O2 Molecular Weight (g/mol): 92.112 Synonym: L-Alanine-3,3,3-d3 (9CI, ACI),L-[3,3,3-d3]Alanine,β,β,β-Trideutero-L-alanine,(2S)-2-Aminopropanoic-3,3,3-d3 acid,L-Alanine-3,3,3-D3,L-Alanine-d3 IUPAC Name: (2S)-2-amino-3,3,3-trideuteriopropanoic acid SMILES: [2H]C([2H])([2H])[C@H](N)C(=O)O