CAS RN 63489-56-5
CAS RN 63489-56-5
IUPAC Name:
4-chloro-3,5,6-trifluoropyridin-2-amine
Synonyms:
4-chloro-3,5,6-trifluoropyridin-2-amine
Molecular Weight (g/mol):
182.53
Molecular Formula:
C5H2ClF3N2
InChi Key:
WZRMLGNDGPCZFG-UHFFFAOYSA-N
SMILES:
NC1=NC(F)=C(F)C(Cl)=C1F
1
–
1
of
1
results
1
2-Amino-4-chloro-3,5,6-trifluoropyridine, 98%
CAS: 63489-56-5 Molecular Formula: C5H2ClF3N2 Molecular Weight (g/mol): 182.53 MDL Number: MFCD08436088 InChI Key: WZRMLGNDGPCZFG-UHFFFAOYSA-N Synonym: 2-amino-4-chloro-3,5,6-trifluoropyridine,4-chloro-3,5,6-trifluoro-2-pyridinamine,4-chloro-3,5,6-trifluoro-2-pyridylamine,3,5,6-trifluoro-4-chloropyridine-2-amine,4-chloranyl-3,5,6-tris fluoranyl pyridin-2-amine PubChem CID: 12508075 IUPAC Name: 4-chloro-3,5,6-trifluoropyridin-2-amine SMILES: C1(=C(C(=C(N=C1N)F)F)Cl)F