CAS RN 6346-09-4
CAS RN 6346-09-4
IUPAC Name:
4,4-diethoxybutan-1-amine
Synonyms:
4,4-diethoxy-butylamine
Molecular Weight (g/mol):
161.25
Molecular Formula:
C8H19NO2
InChi Key:
GFLPSABXBDCMCN-UHFFFAOYSA-N
SMILES:
CCOC(CCCN)OCC
1
–
1
of
1
results
1
4-Aminobutyraldehyde diethyl acetal, 95%
CAS: 6346-09-4 Molecular Formula: C8H19NO2 Molecular Weight (g/mol): 161.24 MDL Number: MFCD00008227 InChI Key: GFLPSABXBDCMCN-UHFFFAOYSA-N IUPAC Name: 4,4-diethoxybutan-1-amine SMILES: CCOC(CCCN)OCC