accessibility menu, dialog, popup

UserName

CAS RN 6342-60-5

CH3ClOO

CAS RN 6342-60-5

IUPAC Name: 2-chloro-5-methylbenzoate
Synonyms: 2-chloro-5-methylbenzoate
Molecular Weight (g/mol): 169.58
Molecular Formula: C8H6ClO2
InChi Key: LEBWXJZAWTVKFL-UHFFFAOYSA-M
SMILES: CC1=CC=C(Cl)C(=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)