accessibility menu, dialog, popup

UserName

CAS RN 6334-18-5

ClOCl

CAS RN 6334-18-5

IUPAC Name: 2,3-dichlorobenzaldehyde
Synonyms: 2,3-dichlorobenzaldehyde
Molecular Weight (g/mol): 175.01
Molecular Formula: C7H4Cl2O
InChi Key: LLMLNAVBOAMOEE-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(C=O)=C1Cl
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)