CAS RN 6331-96-0
CAS RN 6331-96-0
IUPAC Name:
2-amino-4,5-dichlorobenzene-1-sulfonic acid
Synonyms:
2-amino-4,5-dichlorobenzenesulfonic acid
Molecular Weight (g/mol):
242.07
Molecular Formula:
C6H5Cl2NO3S
InChi Key:
AKLDPNVZTZIVFA-UHFFFAOYSA-N
SMILES:
NC1=CC(Cl)=C(Cl)C=C1S(O)(=O)=O
1
–
1
of
1
results
1
2-Amino-4,5-dichlorobenzenesulfonic acid, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality