CAS RN 63245-28-3
CAS RN 63245-28-3
IUPAC Name:
2-[(carboxymethyl)({[(2,6-diethylphenyl)carbamoyl]methyl})amino]acetic acid
Synonyms:
[(carboxymethyl)({[(2,6-diethylphenyl)carbamoyl]methyl})amino]acetic acid
Molecular Weight (g/mol):
322.36
Molecular Formula:
C16H22N2O5
InChi Key:
WNIDXAKKFOKNEF-UHFFFAOYSA-N
SMILES:
CCC1=CC=CC(CC)=C1NC(=O)CN(CC(O)=O)CC(O)=O
1
–
1
of
1
results
1
Etifenin, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.