accessibility menu, dialog, popup

UserName

CAS RN 63226-13-1

H3CNCH3OOOOONCH3CH3

CAS RN 63226-13-1

IUPAC Name: 7-(diethylamino)-3-[7-(diethylamino)-2-oxo-2H-chromene-3-carbonyl]-2H-chromen-2-one
Synonyms: 7-(diethylamino)-3-[7-(diethylamino)-2-oxochromene-3-carbonyl]chromen-2-one
Molecular Weight (g/mol): 460.53
Molecular Formula: C27H28N2O5
InChi Key: SANIRTQDABNCHF-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C=C(C(=O)C3=CC4=CC=C(C=C4OC3=O)N(CC)CC)C(=O)OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results