accessibility menu, dialog, popup

UserName

CAS RN 6321-11-5

ClH3CCH3NH2CH3HO

CAS RN 6321-11-5

IUPAC Name: 4-amino-5-methyl-2-(propan-2-yl)phenol hydrochloridyl
Synonyms: 4-amino-2-isopropyl-5-methylphenol hydrochloridyl
Molecular Weight (g/mol): 200.69
Molecular Formula: C10H15ClNO
InChi Key: YLWORLAQZBRLSS-UHFFFAOYSA-N
SMILES: [Cl].CC(C)C1=CC(N)=C(C)C=C1O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results