accessibility menu, dialog, popup

UserName

CAS RN 632-68-8

KKOIOIOIOOClClClClI

CAS RN 632-68-8

IUPAC Name: dipotassium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Synonyms: dipotassium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Molecular Weight (g/mol): 1049.85
Molecular Formula: C20H2Cl4I4K2O5
InChi Key: VQHHOXOLUXRQFQ-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-]C1=C(I)C=C2C(OC3=C(I)C([O-])=C(I)C=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1I
Color 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)