CAS RN 6310-21-0
CAS RN 6310-21-0
IUPAC Name:
2-tert-butylaniline
Synonyms:
2-tert-butylaniline
Molecular Weight (g/mol):
149.24
Molecular Formula:
C10H15N
InChi Key:
AEIOZWYBDBVCGW-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=CC=C1N