accessibility menu, dialog, popup

UserName

H3CCH3OHOOHO

CAS RN 631-31-2

IUPAC Name: 2-ethyl-2-methylbutanedioic acid
Synonyms: 2-ethyl-2-methyl-succinic acid
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: FDYJJKHDNNVUDR-UHFFFAOYNA-N
SMILES: CCC(C)(CC(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)