accessibility menu, dialog, popup

UserName

CAS RN 631-22-1

CH3OOBrBrOOH3C

CAS RN 631-22-1

IUPAC Name: 1,3-diethyl 2,2-dibromopropanedioate
Synonyms: 1,3-diethyl 2,2-dibromopropanedioate
Molecular Weight (g/mol): 317.96
Molecular Formula: C7H10Br2O4
InChi Key: PFZYFZRUPFUEOB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(Br)(Br)C(=O)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results