CAS RN 6295-87-0
CAS RN 6295-87-0
IUPAC Name:
1-aminopyridin-1-ium iodide
Synonyms:
1-aminopyridin-1-ium iodide
Molecular Weight (g/mol):
222.03
Molecular Formula:
C5H7IN2
InChi Key:
NDRLPYIMWROJBG-UHFFFAOYSA-M
SMILES:
[I-].N[N+]1=CC=CC=C1
1
–
1
of
1
results
1
1-Aminopyridinium iodide, 97%
CAS: 6295-87-0 Molecular Formula: C5H7IN2 Molecular Weight (g/mol): 222.03 MDL Number: MFCD00051893 InChI Key: NDRLPYIMWROJBG-UHFFFAOYSA-M Synonym: 1-aminopyridinium iodide,1-aminopyridin-1-ium iodide,n-aminopyridinium iodide,pyridinium, 1-amino-, iodide,1-aminopyridiniumiodide,pyridylamine, iodide,aminopyridinium iodide,amino-pyridinium iodide,l-aminopyridinium iodide,acmc-209nay PubChem CID: 2733704 IUPAC Name: pyridin-1-ium-1-amine;iodide SMILES: [I-].N[N+]1=CC=CC=C1