CAS RN 6294-17-3
CAS RN 6294-17-3
IUPAC Name:
1-bromo-6-chlorohexane
Synonyms:
1-bromo-6-chlorohexane
Molecular Weight (g/mol):
199.52
Molecular Formula:
C6H12BrCl
InChi Key:
JTYUIAOHIYZBPB-UHFFFAOYSA-N
SMILES:
ClCCCCCCBr
1
–
1
of
1
results
1
1-Bromo-6-chlorohexane, 97%
CAS: 6294-17-3 Molecular Formula: C6H12BrCl Molecular Weight (g/mol): 199.52 MDL Number: MFCD00001019 InChI Key: JTYUIAOHIYZBPB-UHFFFAOYSA-N Synonym: 6-chlorohexyl bromide,hexane, 1-bromo-6-chloro,1-chloro-6-bromohexane,hexyl bromide, 6-chloro,6-bromohexyl chloride,1-bromo-6-chloro hexane,1-bromo-6-chloro-hexane,acmc-1b9m0,1-bromo-6-chlorohexane,jtyuiaohiyzbpb-uhfffaoysa PubChem CID: 80516 IUPAC Name: 1-bromo-6-chlorohexane SMILES: ClCCCCCCBr