accessibility menu, dialog, popup

UserName

NFFFFCl

CAS RN 628692-22-8

IUPAC Name: 2-chloro-3-fluoro-4-(trifluoromethyl)pyridine
Synonyms: 2-chloro-3-fluoro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 199.53
Molecular Formula: C6H2ClF4N
InChi Key: YKQVABFVOGPIKX-UHFFFAOYSA-N
SMILES: FC1=C(C=CN=C1Cl)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results