accessibility menu, dialog, popup

UserName

CAS RN 628-20-6

ClN

CAS RN 628-20-6

IUPAC Name: 4-chlorobutanenitrile
Synonyms: butanenitrile, 4-chloro-
Molecular Weight (g/mol): 103.55
Molecular Formula: C4H6ClN
InChi Key: ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
SMILES: ClCCCC#N
Boiling Point 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)