CAS RN 627-40-7
CAS RN 627-40-7
IUPAC Name:
3-methoxyprop-1-ene
Synonyms:
methallyl alcohol
Molecular Weight (g/mol):
72.11
Molecular Formula:
C4H8O
InChi Key:
FASUFOTUSHAIHG-UHFFFAOYSA-N
SMILES:
COCC=C
1
–
1
of
1
results
1
Allyl methyl ether, 94%
CAS: 627-40-7 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.11 MDL Number: MFCD00008649 InChI Key: FASUFOTUSHAIHG-UHFFFAOYSA-N Synonym: allyl methyl ether,1-propene, 3-methoxy,3-methoxy-1-propene,ether, allyl methyl,3-methoxypropene,methyl allyl ether,allylmethylether,4-oxapent-1-ene,1-methoxy-2-propene,methylallyl ether PubChem CID: 69392 IUPAC Name: 3-methoxyprop-1-ene SMILES: COCC=C