CAS RN 62673-31-8
CAS RN 62673-31-8
IUPAC Name:
bromozincylium; phenylmethanide
Synonyms:
bromozincylium; phenylmethanide
Molecular Weight (g/mol):
236.42
Molecular Formula:
C7H7BrZn
InChi Key:
WRSWIWOVJBYZAW-UHFFFAOYSA-M
SMILES:
[Zn+]Br.[CH2-]C1=CC=CC=C1