accessibility menu, dialog, popup

UserName

CAS RN 62638-05-5

IUPAC Name: strontium(2+) bis(4-cyclohexylbutanoate)
Synonyms: strontium(2+) bis(4-cyclohexylbutanoate)
Molecular Weight (g/mol): 426.11
Molecular Formula: C20H34O4Sr
InChi Key: JBJLAHGTZMLOAH-UHFFFAOYSA-L
SMILES: [Sr++].[O-]C(=O)CCCC1CCCCC1.[O-]C(=O)CCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results