accessibility menu, dialog, popup

UserName

CAS RN 62637-92-7

CH3NCH3NH2CH3NCH3NH2HOOOHO

CAS RN 62637-92-7

IUPAC Name: bis(N1,N1-diethylbenzene-1,4-diamine); oxalic acid
Synonyms: bis(N,N-diethyl-1,4-benzenediamine); oxalic acid
Molecular Weight (g/mol): 418.54
Molecular Formula: C22H34N4O4
InChi Key: JJEXNPALAITIMN-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.CCN(CC)C1=CC=C(N)C=C1.CCN(CC)C1=CC=C(N)C=C1
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)