accessibility menu, dialog, popup

UserName

CAS RN 6228-73-5

IUPAC Name: cyclopropanecarboxamide
Synonyms: cyclopropanecarboxamide
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: AIMMVWOEOZMVMS-UHFFFAOYSA-N
SMILES: NC(=O)C1CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results