CAS RN 622387-85-3
CAS RN 622387-85-3
IUPAC Name:
3-[(1-methyl-1H-indol-3-yl)methylidene]-2-oxo-2,3-dihydro-1H-indole-5-sulfonamide
Synonyms:
3-[(1-methylindol-3-yl)methylidene]-2-oxo-1H-indole-5-sulfonamide
Molecular Weight (g/mol):
353.4
Molecular Formula:
C18H15N3O3S
InChi Key:
MLKHXLFEYOOYEY-UHFFFAOYSA-N
SMILES:
CN1C=C(C=C2C(=O)NC3=C2C=C(C=C3)S(N)(=O)=O)C2=CC=CC=C12
1
–
1
of
1
results
1
Thermo Scientific™ Syk Inhibitor
CAS: 622387-85-3 Molecular Formula: C18H15N3O3S Synonym: Spleen Tyrosine Kinase Inhibitor