CAS RN 622-08-2
CAS RN 622-08-2
IUPAC Name:
2-(benzyloxy)ethan-1-ol
Synonyms:
benzyl glycol
Molecular Weight (g/mol):
152.19
Molecular Formula:
C9H12O2
InChi Key:
CUZKCNWZBXLAJX-UHFFFAOYSA-N
SMILES:
OCCOCC1=CC=CC=C1
1
–
1
of
1
results
1
2-Benzyloxyethanol, 98%
CAS: 622-08-2 Molecular Formula: C9H12O2 Molecular Weight (g/mol): 152.19 MDL Number: MFCD00002868 InChI Key: CUZKCNWZBXLAJX-UHFFFAOYSA-N Synonym: 2-benzyloxy ethanol,2-benzyloxyethanol,ethanol, 2-phenylmethoxy,ethylene glycol monobenzyl ether,2-benzyloxy-1-ethanol,benzyl cellosolve,glycol benzyl ether,ethanol, 2-benzyloxy,glycol monobenzyl ether PubChem CID: 12141 IUPAC Name: 2-phenylmethoxyethanol SMILES: OCCOCC1=CC=CC=C1