CAS RN 619-33-0
IUPAC Name:
1,1-dichloro-2,2-diethoxyethane
Synonyms:
1,1-dichloro-2,2-diethoxyethane
Molecular Weight (g/mol):
187.06
Molecular Formula:
C6H12Cl2O2
InChi Key:
CDHLQZJRWKQATP-UHFFFAOYSA-N
SMILES:
CCOC(OCC)C(Cl)Cl
1
–
1
of
1
results
1
Dichloroacetaldehyde Diethyl Acetal, TRC
CAS: 619-33-0 Molecular Formula: C6 H12 Cl2 O2 Molecular Weight (g/mol): 187.064 Synonym: 1,1-Dichloro-2,2-diethoxy-ethane,Dichloro-acetaldehyde Diethyl Acetal,1,1-Dichloro-2,2-diethoxyethane,2,2-Dichloro-1,1-diethoxyethane,Dichloroacetaldehyde Diethyl Acetal IUPAC Name: 1,1-dichloro-2,2-diethoxyethane SMILES: CCOC(OCC)C(Cl)Cl