accessibility menu, dialog, popup

UserName

HOIOOO

CAS RN 61717-82-6

IUPAC Name: 1-hydroxy-1-oxo-3H-1λ⁵,2-benziodaoxol-3-one
Synonyms: 2-iodoxybenzoic acid
Molecular Weight (g/mol): 280.02
Molecular Formula: C7H5IO4
InChi Key: CQMJEZQEVXQEJB-UHFFFAOYSA-N
SMILES: O[I]1(=O)OC(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)