accessibility menu, dialog, popup

UserName

CAS RN 61652-81-1

H3CSClOO

CAS RN 61652-81-1

IUPAC Name: decane-1-sulfonyl chloride
Synonyms: decane-1-sulfonyl chloride
Molecular Weight (g/mol): 240.79
Molecular Formula: C10H21ClO2S
InChi Key: WCKKWABZCCKDTN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCS(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results