accessibility menu, dialog, popup

UserName

CAS RN 616-42-2

CH3OSOOH3C

CAS RN 616-42-2

IUPAC Name: dimethyl sulfite
Synonyms: dimethyl sulfite dimethylsulfit
Molecular Weight (g/mol): 110.13
Molecular Formula: C2H6O3S
InChi Key: BDUPRNVPXOHWIL-UHFFFAOYSA-N
SMILES: COS(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results