accessibility menu, dialog, popup

UserName

CAS RN 615-22-5

H3CSSN

CAS RN 615-22-5

IUPAC Name: 2-(methylsulfanyl)-1,3-benzothiazole
Synonyms: 2-(methylthio)benzothiazole MTBT benzothiazolethiol, 2-methyl-
Molecular Weight (g/mol): 181.27
Molecular Formula: C8H7NS2
InChi Key: UTBVIMLZIRIFFR-UHFFFAOYSA-N
SMILES: CSC1=NC2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)